Would you like to log in again?
Exocell The Creatinine Companion, Creatinine ELISA Assay, Ethos Biosciences
Supplier: Ethos Biosciences
1012
MSPP-1012SPEA 636.92 USD InStock
MSPP-1012SP
Exocell The Creatinine Companion, Creatinine ELISA Assay, Ethos Biosciences
Assays ELISAs
Chemical Assay
  • Specimen required: Urine, 20 µL
Precision: Intrassay and interassay precision for samples within useful range of the assay have a C.V. ≦ 10% of the mean
2 x 96 well plates. 88 sample wells per plate. 8 standard wells per plate. 176 total sample wells. 176 total samples or 88 in duplicate (recommended).
Ordering information: Our kits provide all of the necessary components, including overages, to conduct your assays with confidence.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty